Order Entry
Belgium
ContactUsLinkComponent
ReadiLink™ xtra Rapid XFD647 Antibody Labelling Kit *BSA-Compatible, XFD647 Same Structure to Alexa Fluor™ 647* (2 Labelings)
ReadiLink™ xtra Rapid XFD647 Antibody Labelling Kit *BSA-Compatible, XFD647 Same Structure to Alexa Fluor™ 647* (2 Labelings)
Catalog # AATB1985
Supplier:  AAT Bioquest
CAS Number:  
undefined
ReadiLink™ xtra Rapid XFD647 Antibody Labelling Kit *BSA-Compatible, XFD647 Same Structure to Alexa Fluor™ 647* (2 Labelings)
Catalog # AATB1985
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    2 Tests
  • Description:
    ReadiLink™ xtra rapid XFD647 antibody labelling kit 'BSA-compatible, XFD647 same structure to Alexa Fluor™ 647'

Specifications

About this item

XFD647 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 647 (Alexa Fluor® is the trademark of ThermoFisher).

  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labeling protocol
  • No column purification required
Caution: For research use only.