ContactUsLinkComponent
Amplite® Colorimetric Butyrylcholinesterase Activity Assay Kit
Amplite® Colorimetric Butyrylcholinesterase Activity Assay Kit
Catalog # AATB11406
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Colorimetric Butyrylcholinesterase Activity Assay Kit
Catalog # AATB11406
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Amplite™ colorimetric butyrylcholinesterase activity assay kit
Specifications
About this item
Butyrylcholinesterase (EC 3.1.1.8; BChE), also known as pseudocholinesterase or plasma cholinesterase, is mainly synthesised in liver and present in blood.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.