ContactUsLinkComponent
ReadiLink™ iFluor® 488 FISH Fluorescence Imaging Kit
ReadiLink™ iFluor® 488 FISH Fluorescence Imaging Kit
Catalog # AATB17310
Supplier:  AAT Bioquest
CAS Number:  
undefined
ReadiLink™ iFluor® 488 FISH Fluorescence Imaging Kit
Catalog # AATB17310
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    25 Tests
  • Description:
    Convenient tool for labelling a target DNA using an iFluor® 488 labeled FISH probe via in situ hybridization, for PCR microplates
Specifications
About this item
The kit provides Taq DNA polymerase enzyme, which incorporates iFluor® 488-dUTPs in the target DNA through Polymerase Chain Reaction (PCR).
  • Robust protocol
  • Bright labelling
  • Photostable fluorescence
The Readilink™ iFluor® 488 FISH fluorescence imaging kit is a convenient tool for labelling a target DNA using an iFluor® 488 labeled FISH probe via in situ hybridisation.
Our proprietary iFluor® dyes are brighter and more photostable than traditional fluorescent labels, providing the desired resolution and signal.
Packaging: Packaged in plastic vials.
Caution: For research use only.