ReadiLink™ iFluor® 488 Oligo and ssDNA Labelling Kit
Supplier: AAT Bioquest
Certificates
About this item
The kit is optimised for efficient labelling and contains all the essential reagents required for efficient labelling of ssDNA or oligos.
- Efficient labelling
- Contains all essential reagents
- Multiple applications
The resulting iFluor® 488-labeled DNA probes are ideally suited for biological applications, e.g., electrophoretic mobility shift assays (EMSA), Northern and Southern blots, colony or in situ hybridisations..
Packaging: Packaged in plastic vials.
Caution: For research use only.