Order Entry
Belgium
ContactUsLinkComponent
Cell Meter™ Fluorimetric Live Cell Cycle Assay Kit *Optimised for 405 nm Violet Laser Excitation*
Cell Meter™ Fluorimetric Live Cell Cycle Assay Kit *Optimised for 405 nm Violet Laser Excitation*
Catalog # AATB22845
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Fluorimetric Live Cell Cycle Assay Kit *Optimised for 405 nm Violet Laser Excitation*
Catalog # AATB22845
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ Fluorimetric live cell cycle assay kit *Optimized for 405 nm violet laser excitation*

Specifications

About this item

The cell cycle has four sequential phases: G0/G1, S, G2, and M.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Optimised for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.