Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
Catalog # AATB22326
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:200 Tests
Specifications
About this item
It is sensitive and might be used for measuring active DNA synthesis at single-cell level.
- FITC filter set compatible
- Bright fluorescence
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit uses XdU which is incorporated into cellular DNA during DNA synthesis. After fixing cells, the incorporated XdU is labelled with iFluor® 488 MTA. The resulted iFluor® 488-labeled DNA formed in cells is imaged with a FITC filter set. Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry-based assay..
Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay.