Order Entry
Belgium
ContactUsLinkComponent
Cell Meter™ Cell Viability Assay Kit *NIR Fluorescence Optimised for Fluorescence Microplate Reader*
Cell Meter™ Cell Viability Assay Kit *NIR Fluorescence Optimised for Fluorescence Microplate Reader*
Catalog # AATB22787
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Cell Viability Assay Kit *NIR Fluorescence Optimised for Fluorescence Microplate Reader*
Catalog # AATB22787
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Description:
    Cell Meter™ cell viability assay kit *NIR fluorescence optimized for fluorescence microplate reader*

Specifications

About this item

Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
  • Results in under 2 hours
Caution: For research use only.