Order Entry
Belgium
ContactUsLinkComponent
 
Cell Meter™ Nuclear Apoptosis Assay Kit *Green Fluorescence Optimised for Flow Cytometry*
Catalog # AATB22811
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Nuclear Apoptosis Assay Kit *Green Fluorescence Optimised for Flow Cytometry*
Catalog # AATB22811
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ nuclear apoptosis assay kit *green fluorescence optimized for flow cytometry*

Specifications

About this item

Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Optimised for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.