Cell Meter™ Caspase 8 Activity Apoptosis Assay Kit *Green Fluorescence*
Catalog # AATB22798
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:200 Tests
- Description:Cell Meter™ Caspase 8 activity apoptosis assay kit *green fluorescence*
Specifications
About this item
The Cell Meter™ assay kits are a set of tools for monitoring cellular functions.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
Caution: For research use only.