Order Entry
Belgium
ContactUsLinkComponent
Cell Meter™ Fixed Cell and Tissue TUNEL Apoptosis Assay Kit *Green Fluorescence*
Cell Meter™ Fixed Cell and Tissue TUNEL Apoptosis Assay Kit *Green Fluorescence*
Catalog # AATB22851
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Fixed Cell and Tissue TUNEL Apoptosis Assay Kit *Green Fluorescence*
Catalog # AATB22851
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    25 Tests
  • Description:
    Cell Meter™ fixed cell and tissue tunel apoptosis assay kit *green fluorescence*

Specifications

About this item

Cell Meter™ fixed cell and tissue TUNEL apoptosis assay kit provides a robust tool for conveniently detecting DNA fragmentation caused by apoptosis.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.