Orders LinkContactUsLinkComponent
Molecular models: crystal structures
Molecular models: crystal structures
Marketsource
Supplier:   SPIRING ENTERPRISES
Molecular models: crystal structures
Marketsource
Certificates
Molecular models: crystal structures
Molecular models: crystal structures
5 Options Available Below
Product Details & Documents
Finished sets with selected atoms and bonds to show different types of crystal structures.
Delivery information: Supplied assembled or with instructions for easy assembly.
Product Family Options
Crystals with covalent bonding
Product Information
  • Availability
    Price
  • 764-0069
    Molecular model of sphalerite
    Molecular model of sphalerite
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 22,61 €
    22,61 €
    1 * 1 SET
       22,61 €
    Specifications:
    Marketsource
    Marketsource
  • 764-0074
    Molecular model of sulphur
    Molecular model of sulphur
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 12,91 €
    12,91 €
    1 * 1 SET
       12,91 €
    Specifications:
    Marketsource
    Marketsource
  • 764-0066
    Molecular model of Buckminster Fullerene
    Molecular model of Buckminster Fullerene
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,87 €
    25,87 €
    1 * 1 ST
       25,87 €
    Specifications:
    Marketsource
    Marketsource
  • 764-0067
    Molecular model of ice
    Molecular model of ice
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,60 €
    24,60 €
    1 * 1 ST
       24,60 €
    Specifications:
    Marketsource
    Marketsource
Crystals with ionic bonding – Salts
Product Information
  • Availability
    Price
  • 764-0071
    Molecular model of calcium fluoride, 30 atoms
    Molecular model of calcium fluoride, 30 atoms
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
    Specifications:
    Marketsource
    Marketsource