Order Entry
Belgium
ContactUsLinkComponent
ModDetect™ Phosphorothioate Panel
ModDetect™ Phosphorothioate Panel
Catalog # ROCKKNA-100
CAS Number:  
ModDetect™ Phosphorothioate Panel
Catalog # ROCKKNA-100
CAS Number:  

Specifications

  • Pack type:
    Combo pack
  • Pk:
    1 SET
  • Description:
    ModDetect™ phosphorothioate panel
  • Storage conditions:
    Dry Ice
  • Storage temperature:
    Store five phosphorothioate vials at −20 °C or below prior to opening. Store the vial at −20 °C or below after dilution. Avoid cycles of freezing and thawing.
  • Format:
    Liquid and Lyophilised

Specifications

About this item

The set includes five unlabelled ModDetect™ phosphorothioate detection reagents, curated by binding characteristics, and three reporter molecules labelled with peroxidase, 488 λ fluorophore and 649 λ fluorophore.

The different binding characteristics create opportunities to generate highly optimised immunoassays independent of oligonucleotide sequence, as opposed to spending significant efforts to optimise assays to underperforming reagents.

Designed for use in various immunoassays, such as ELISA, IF, and IHC.

Store three reporter molecule vials at 4 °C or −20 °C prior to restoration. For extended storage aliquot contents and freeze at −20 °C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.

Expiration date is one year from date of receipt.

This set includes the following components: PS03, PS04, PS05, PS07, PS09, and three reporter molecules.