Order Entry
Belgium
ContactUsLinkComponent
 
MegaWox™ polyHRP Streptavidin Conjugate (1 mg)
Catalog # AATB11039
Supplier:  AAT Bioquest
CAS Number:  
undefined
MegaWox™ polyHRP Streptavidin Conjugate (1 mg)
Catalog # AATB11039
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 mg
  • Conjugation:
    Streptavidin
  • Source:
    Eukaryotic cells

Specifications

About this item

Immunoassays (such as histochemistry, ELISAs and Western blotting) are important in clinical diagnosis and experimental protein analysis.

  • High sensitivity
  • Suitable for multiple applications
  • Highly purified
Caution: For research use only.