ContactUsLinkComponent
FluoroQuest™ Anti-Fading Mounting Medium with DAPI (20 ml)
  AATB20005
 :  
undefined
FluoroQuest™ Anti-Fading Mounting Medium with DAPI (20 ml)
  AATB20005
 :  
  • Pk:
    20 mL
  • Description:
    FluoroQuest™ Anti-fading Mounting Medium with DAPI
FluoroQuest™ with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.
  • Preserves fluorescence
  • Prevents photobleaching
  • Contains nuclear counterstain for convenience
 : For research use only.