Order Entry
Belgium
Orders LinkContactUsLinkComponent
FluoroQuest™ Fluorescence Signal Enhancing Solution (5 ml)

FluoroQuest™ Fluorescence Signal Enhancing Solution (5 ml)

FluoroQuest™ fluorescence signal enhancing solution, 5 mL
Catalog # AATB20006
Supplier:   AAT Bioquest
FluoroQuest™ Fluorescence Signal Enhancing Solution (5 ml)
Catalog # AATB20006
Supplier:  AAT Bioquest
CAS Number:  
FluoroQuest™ Fluorescence Signal Enhancing Solution (5 ml)
FluoroQuest™ Fluorescence Signal Enhancing Solution (5 ml)

FluoroQuest™ fluorescence signal enhancing solution, 5 mL

CAS Number:  
Price
156,00 €
Log in for availability
1 * 5 mL
Product Details & Documents

When proteins are conjugated to fluorescent organic dyes, fluorescence emission of the dye molecules is usually decreased, sometimes up to 50 to 70%.

  • Significantly improved signal intensity
  • Suitable for all fluorescence platforms
  • Multiple applications

Caution: For research use only.

Specifications

Specifications

  • Pk:
    5 mL
  • Description:
    FluoroQuest™ fluorescence signal enhancing solution

Specifications