Order Entry
Belgium
ContactUsLinkComponent
Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), Spectinomycin
Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), Spectinomycin
Catalog # AATB50039
Supplier:  AAT Bioquest
CAS Number:  
undefined
Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), Spectinomycin
Catalog # AATB50039
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 mg
  • Conjugation:
    Spectinomycin
  • Protein/peptide type:
    Native
  • Source:
    Serum
  • Species:
    Megathura crenulata
  • Protein synonyms:
    KLH
  • Protein/peptide name:
    Keyhole limpet hemocyanin

Specifications

About this item

Spectinomycin-KLH conjugate is used for developing anti-spectinomycin antibodies that are typically used in an ELISA assay.

  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Caution: For research use only.