Native Human Concanavalin A (from Human), Alexa Fluor® 647
Catalog # AATB25595
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Vial
- Pk:1 mg
- Conjugation:Alexa Fluor® 647
- Protein/peptide type:Native
- Species:Human
- Protein synonyms:ConA
- Protein/peptide name:Concanavalin A
Specifications
About this item
Concanavalin A (ConA) is a lectin that binds specifically to certain structures found in various sugars, glycoproteins and glycolipids.
- Well-cited conjugate
- High affinity and selectivity
- Multiple applications
Caution: For research use only.