Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), mFluor™ Red 780
Catalog # AATB16948
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 µG
- Conjugation:mFluor™ Red 780
- Protein/peptide type:Native
- Source:Streptomyces avidinii
- Species:Streptomyces avidinii
- Protein synonyms:Avidin
- Protein/peptide name:Streptavidin
Specifications
About this item
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.
- Optimized for flow cytometry
- Useful for assay development
- High affinity
- Multiple applications
Caution: For research use only.