Native Amanita phalloides Phalloidin (from Eukaryotic cells), Texas Red®
Catalog # AATB23103
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Vial
- Pk:300 Tests
- Conjugation:Texas Red®
- Protein/peptide type:Native
- Source:Eukaryotic cells
- Species:Amanita phalloides
- Protein synonyms:Amanita phalloides
- Protein/peptide name:Phalloidin
Specifications
About this item
This red fluorescent phalloidin conjugate (equivalent to Texas Red®-labeled phalloindin) selectively binds to F-actins with much higher photostability than the fluorescein-phalloidin conjugates.
- Suitable for all fluorescent imaging platforms
- Specific for actin in both living or fixed cells
- Multiple applications
Caution: For research use only.