Order Entry
Belgium
ContactUsLinkComponent
Native 2',3'-cGAMP (from Mammalian cells), Biotin
Native 2',3'-cGAMP (from Mammalian cells), Biotin
  AATB20316
 :  
undefined
Native 2',3'-cGAMP (from Mammalian cells), Biotin
  AATB20316
 :  

 

  • Pack type:
    Vial
  • Pk:
    100 µG
  • Conjugation:
    Biotin
  • Protein/peptide type:
    Native
  • Source:
    Mammalian cells
  • Species:
  • Environmentally Preferable:
  • Protein synonyms:
    Cyclic guanosine monophosphate adenosine monophosphate|cGAMP
  • Protein/peptide name:
    2',3'-cGAMP

 

 

2',3'-cGAMP or cGAMP (Cyclic guanosine monophosphate 'adenosine monophosphate) is produced in mammalian cells by cGAS (cGAMP synthase) from ATP and GTP upon cytosolic DNA stimulation.

  • Essential component of intracellular signaling
 : For research use only.