Order Entry
Belgium
ContactUsLinkComponent
 
Native Streptavidin (from Streptomyces avidinii), Alexa Fluor® 546
  AATB16958
 :  
undefined
Native Streptavidin (from Streptomyces avidinii), Alexa Fluor® 546
  AATB16958
 :  

 

  • Pack type:
    Vial
  • Pk:
    200 µG
  • Conjugation:
    Alexa Fluor® 546
  • Protein/peptide type:
    Native
  • Source:
    Streptomyces avidinii
  • Protein synonyms:
    Avidin|Streptomyces avidinii
  • Protein/peptide name:
    Streptavidin

 

 

Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.

  • Bright signal
  • More photostable fluorescence
  • Improved proprietary fluorescent label
 : For research use only.