Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ac-DEVD-AMC

Ac-DEVD-AMC

Plastic Bottle, 5 mg
Catalog # AATB13402
Supplier:   AAT Bioquest
Ac-DEVD-AMC
Catalog # AATB13402
Supplier:  AAT Bioquest
CAS Number:  
Ac-DEVD-AMC
Ac-DEVD-AMC

Plastic Bottle, 5 mg

CAS Number:  
Price
156,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

Ac-DEVD-AMC is a fluorogenic substrate for caspase 3, a protease that is rapidly activated when cells are exposed to apoptotic conditions and that cleaves poly(ADP-ribose) polymerase.

  • Hazard Class/ Packing Group: 5, II
  • GHS symbol: GHS07
  • Signal Word: Warning
Specifications

Specifications

  • Pack type:
    Plastic Bottle
  • Pk:
    5 mg
  • Cat. No.:
    AATB13402
  • Protein/peptide name:
    Ac-DEVD-AMC
  • Purity:
    90%

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.