Order Entry
Belgium
ContactUsLinkComponent
DY677 CEP (DyLight 677 Phosphoramidite)
DY677 CEP (DyLight 677 Phosphoramidite)
Catalog # AATB6078
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
DY677 CEP (DyLight 677 Phosphoramidite)
Catalog # AATB6078
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    100 µmol
  • Host:
  • Description:
    For efficiently labeling an oligonucleotide
  • Environmentally Preferable:

Specifications

About this item

DY677 CEP (DyLight 677 phosphoramidite) is an excellent replacement to the commonly used Cy5.5 phosphoramidite.

  • High stability
  • Oligo deprotection can be carried out at room temperature

DY677 CEP appears to be more stable than Cy5.5 phosphoramidite. As Cy5.5 phosphoramidite, the deprotection of oligos containing DY677 CEP may be carried out with ammonium hydroxide at room temperature, regardless of the base protecting groups on the monomers used.

It is always prudent to use monomers with base labile protecting groups to limit the exposure time to 2 hours or less at 65 °C during deprotection.

Packaging: Plastic vial.
Caution: For research use only.