Order Entry
Belgium
ContactUsLinkComponent
ReadiLink™ Rapid XFD594 Antibody Labelling Kit 'XFD594 Same Structure to Alexa Fluor™ 594' (2×50 µg Labellings)
ReadiLink™ Rapid XFD594 Antibody Labelling Kit 'XFD594 Same Structure to Alexa Fluor™ 594' (2×50 µg Labellings)
Catalog # AATB1277
Supplier:  AAT Bioquest
CAS Number:  
undefined
ReadiLink™ Rapid XFD594 Antibody Labelling Kit 'XFD594 Same Structure to Alexa Fluor™ 594' (2×50 µg Labellings)
Catalog # AATB1277
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Description:
    ReadiLink™ rapid XFD594 antibody labeling kit 'XFD594 same structure to Alexa Fluor™ 594'

Specifications

About this item

XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).

  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labelling protocol
  • No column purification required
Caution: For research use only.