Order Entry
Belgium
ContactUsLinkComponent
FluoroQuest™ Fluorescence Quantum Yield Determination Kit *Optimized for Bioconjugates* (1 kit)
FluoroQuest™ Fluorescence Quantum Yield Determination Kit *Optimized for Bioconjugates* (1 kit)
Catalog # AATB20
Supplier:  AAT Bioquest
CAS Number:  
undefined
FluoroQuest™ Fluorescence Quantum Yield Determination Kit *Optimized for Bioconjugates* (1 kit)
Catalog # AATB20
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    FluoroQuest™ fluorescence quantum yield determination kit

Specifications

About this item

When a fluorophore absorbs a photon of light, an energetically excited state is formed.

  • Robust protocol
  • Contains all necessary components
Caution: For research use only.