ContactUsLinkComponent
ReadiLink™ Rapid iFluor® 350 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
ReadiLink™ Rapid iFluor® 350 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Catalog # AATB1220
Supplier:  AAT Bioquest
CAS Number:  
undefined
ReadiLink™ Rapid iFluor® 350 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Catalog # AATB1220
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    2 Tests
  • Description:
    ReadiLink™ rapid iFluor™ 350 antibody labeling kit 'Microscale optimized for labeling 50 µg antibody per reaction'
Specifications
About this item
AAT Bioquest's iFluor® dyes are optimised for labelling proteins, in particular, antibodies.
  • Violet-laser excitable
  • Robust and simple antibody labelling protocol
  • No column purification required
Caution: For research use only.