Order Entry
Belgium
ContactUsLinkComponent
ReadiLink™ xtra Rapid Cy5 Antibody Labelling Kit *BSA-Compatible* (2 Labellings)
ReadiLink™ xtra Rapid Cy5 Antibody Labelling Kit *BSA-Compatible* (2 Labellings)
Catalog # AATB1972
Supplier:  AAT Bioquest
CAS Number:  
undefined
ReadiLink™ xtra Rapid Cy5 Antibody Labelling Kit *BSA-Compatible* (2 Labellings)
Catalog # AATB1972
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    2 Tests
  • Description:
    ReadiLink™ xtra rapid Cy5 antibody labeling kit 'BSA-compatible'

Specifications

About this item

ReadiLink™ xtra rapid antibody labelling kits require essentially only 2 simple mixing steps without a column purification needed.

  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labelling protocol
  • No column purification required
Caution: For research use only.