ReadiLink™ xtra Rapid iFluor® 555 Antibody Labelling Kit *BSA-Compatible* (2 Labellings)
Catalog # AATB1958
Fournisseur: AAT Bioquest
Numéro CAS:
Spécifications
- Cdt:2 Tests
- Désignation:ReadiLink™ xtra rapid iFluor™ 555 antibody labeling kit 'BSA-compatible'
Spécifications
À propos de cet article
ReadiLink™ xtra rapid antibody labelling kits require essentially only two simple mixing steps without a column purification needed.
- Contains all essential components
- Minimal hands on time
- Ready to use
- Robust and simple antibody labelling protocol
- No column purification required
Attention: For research use only.