ReadiLink™ Rapid FITC Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Catalog # AATB1299
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:2 Tests
- Description:ReadiLink™ rapid FITC antibody labeling kit 'Microscale optimized for labeling 50 µg antibody per reaction'
Specifications
About this item
FITC is still the most popular fluorescent labelling dye for preparing green fluorescent bioconjugates.
- Contains all essential components
- Minimal hands on time
- Ready to use
- Robust and simple antibody labelling protocol
- No column purification required
Caution: For research use only.