Order Entry
Belgium
ContactUsLinkComponent
Portelite™ Fluorimetric ssDNA Quantitation Kit
Portelite™ Fluorimetric ssDNA Quantitation Kit
Catalog # AATB17625
Supplier:  AAT Bioquest
CAS Number:  
undefined
Portelite™ Fluorimetric ssDNA Quantitation Kit
Catalog # AATB17625
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Description:
    Kit is designed to rapidly measure single-stranded DNA

Specifications

About this item

The assay detects long oligonucleotides or ssDNA. Nucleotides and short oligo nucleotides of six bases or less do not interfere in the quantitation assay.

  • Optimized for cytocite and qubit fluorometers
  • Any volume from 1 to 20 µl is acceptable
  • Assay range of 1 to 200 ng

The kit has all the essential reagents, including Helixyte™ Green ssDNA reagent, dilution buffer, and prediluted DNA standards. Helixyte™ Green ssDNA reagent is a sensitive fluorescent nucleic acid probe for quantifying oligonucleotides and single-stranded DNA (ssDNA) in solution.

Packaging: Plastic vial.
Caution: For research use only.