ContactUsLinkComponent
Helixyte™ Green Fluorometric dsDNA Quantitation kit *High Sensitivity*
Helixyte™ Green Fluorometric dsDNA Quantitation kit *High Sensitivity*
Catalogus # AATB17651
Leverancier:  AAT BIOQUEST
CAS-nummer:  
undefined
Helixyte™ Green Fluorometric dsDNA Quantitation kit *High Sensitivity*
Catalogus # AATB17651
Leverancier:  AAT BIOQUEST
CAS-nummer:  
Specificaties
  • Pk:
    200 Tests
  • Description:
    Helixyte™ green fluorimetric dsDNA quantitation kit *high sensitivity*
Specificaties
Over dit artikel
Helixyte™ Green dsDNA Quantitation Assay Kit can be used for selectively detecting as little as 25 pg/ml of dsDNA in the presence of ssDNA, RNA, and free nucleotides.
  • High sensitivity
  • Exceptional DNA affinity
  • Robust protocol
  • Multiple applications
Let op: For research use only.