ContactUsLinkComponent
Portelite™ Fluorimetric DNA Quantitation Kit with Broad Dynamic Range *Optimized for Cytocite™ and Qubit™ Fluorometers*
Portelite™ Fluorimetric DNA Quantitation Kit with Broad Dynamic Range *Optimized for Cytocite™ and Qubit™ Fluorometers*
Catalog # AATB17665
Supplier:  AAT Bioquest
CAS Number:  
undefined
Portelite™ Fluorimetric DNA Quantitation Kit with Broad Dynamic Range *Optimized for Cytocite™ and Qubit™ Fluorometers*
Catalog # AATB17665
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Portelite™ fluorimetric DNA quantitation kit with broad dynamic range *optimized for cytocite™ and qubit™ fluorometers*
Specifications
About this item
DNA Quantitation is a very important task in DNA sample preparations for various analyses.
  • Linear over three orders of magnitude
  • Exceptional dsDNA selectivity over rna
  • Robust protocol
  • Multiple applications
Caution: For research use only.