ContactUsLinkComponent
Signal Guard™ HRP Reaction Stopping Solution
  AATB11020
 :  
undefined
Signal Guard™ HRP Reaction Stopping Solution
  AATB11020
 :  
  • Pk:
    500 Tests
  • Description:
    Signal Guard™ HRP reaction stopping solution
HRP coupling reactions provide sensitive biomolecular assays based on hydrogen peroxide- generating enzyme systems linked to peroxidase-mediated oxidation.
  • Improves assay sensitivity and control
  • Easy to use
  • Adaptable to multiple hrp reaction systems
 : For research use only.