ContactUsLinkComponent
Signal Guard™ HRP Conjugate Stabilizer
Signal Guard™ HRP Conjugate Stabilizer
Catalog # AATB11010
Supplier:  AAT Bioquest
CAS Number:  
undefined
Signal Guard™ HRP Conjugate Stabilizer
Catalog # AATB11010
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    50 mL
  • Description:
    Signal Guard™ HRP conjugate stabilizer
Specifications
About this item
The major advantage of a liquid HRP-conjugate is that it eliminates the chance for human error when dissolving or diluting the lyophilized or concentrated conjugate.
  • Permits liquid-hrp conjugates to be stored at 4°c for 24 months or RT for 6 months
  • Can create stock solutions or ready-to-use reagents
Caution: For research use only.