ContactUsLinkComponent
Amplite® MMP-3 Activity Assay Kit *Green Fluorescence*
Amplite® MMP-3 Activity Assay Kit *Green Fluorescence*
Catalog # AATB13512
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® MMP-3 Activity Assay Kit *Green Fluorescence*
Catalog # AATB13512
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Amplite™ MMP-3 activity assay kit *green fluorescence*
  • Tests per kit:
    100 Tests
Specifications
About this item
The matrix metalloproteinases (MMPs) constitute a family of zinc-dependent endopeptidases that function within the extracellular matrix.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.