Saisie de commande
Belgium
ContactUsLinkComponent
Amplite® Fluorimetric Xanthine Oxidase Assay Kit *Red Fluorescence*
Amplite® Fluorimetric Xanthine Oxidase Assay Kit *Red Fluorescence*
Catalog # AATB11304
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Fluorimetric Xanthine Oxidase Assay Kit *Red Fluorescence*
Catalog # AATB11304
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Cdt:
    200 Tests
  • Désignation:
    Amplite™ Fluorimetric xanthine oxidase assay kit *red fluorescence*
  • Tests par set:
    200 Tests
Specifications
About this item
Xanthine oxidase (XO) is an enzyme that catalyses the oxidation of hypoxanthine to xanthine and can further catalyze the oxidation of xanthine to uric acid.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.