ContactUsLinkComponent
Amplite® Fluorimetric Catalase Assay Kit *Red Fluorescence*
Amplite® Fluorimetric Catalase Assay Kit *Red Fluorescence*
Catalog # AATB11306
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Fluorimetric Catalase Assay Kit *Red Fluorescence*
Catalog # AATB11306
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ fluorimetric catalase assay kit *red fluorescence*
  • Tests per kit:
    200 Tests
Specifications
About this item
Catalase is a common antioxidant heme-containing redox enzyme found in nearly all living organisms that are exposed to oxygen.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.