ContactUsLinkComponent
Amplite® Colorimetric Enterokinase Activity Assay Kit
Amplite® Colorimetric Enterokinase Activity Assay Kit
Catalog # AATB11410
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Colorimetric Enterokinase Activity Assay Kit
Catalog # AATB11410
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ colorimetric enterokinase activity assay kit
  • Tests per kit:
    200 Tests
Specifications
About this item
Enterokinase (also called enteropeptidase) is a serine protease produced by cells in the duodenal wall and is a key enzyme in human and animal digestion system.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.