Cell Meter™ Caspase 3/7 Activity Apoptosis Assay Kit *Green Fluorescence Optimized for Flow Cytometry*
N° de catalogue AATB22823
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:100 Tests
- Description:Cell Meter™ Caspase 3/7 activity apoptosis assay kit *green fluorescence optimized for flow Cytometry*
- Tests per kit:100 Tests
Specifications
About this item
Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Optimized for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.