Order Entry
Belgium
ContactUsLinkComponent
Cell Explorer™ Live Cell Tracking Kit *Orange Fluorescence*
Cell Explorer™ Live Cell Tracking Kit *Orange Fluorescence*
Catalog # AATB22622
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Explorer™ Live Cell Tracking Kit *Orange Fluorescence*
Catalog # AATB22622
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Description:
    Cell Explorer™ Live cell tracking kit *orange fluorescence*
  • Tests per kit:
    200 Tests

Specifications

About this item

Our Cell Explorer™ Live Cell Tracking Kit uses a proprietary orange fluorescent dye that gets enhanced fluorescence upon entering into live cells.

  • Long label retention
  • Suitable for all major fluorescence platforms
  • Contains all necessary components
  • Multiple applications
Caution: For research use only.