Order Entry
Belgium
ContactUsLinkComponent
Cell Meter™ Fluorescence Gap Junction Tracing Kit
Cell Meter™ Fluorescence Gap Junction Tracing Kit
Catalog # AATB23600
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Fluorescence Gap Junction Tracing Kit
Catalog # AATB23600
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ Fluorescence gap junction tracing kit
  • Tests per kit:
    100 Tests

Specifications

About this item

Gap junctions are specialised intercellular membrane channels constituted with connexin that selectively facilitate the passage of small molecules of <1,5 Kd across cells.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.