Order Entry
Belgium
ContactUsLinkComponent
Live or Dead™ Fixable Dead Cell Staining Kit *Red Fluorescence Optimized for Flow Cytometry*
Live or Dead™ Fixable Dead Cell Staining Kit *Red Fluorescence Optimized for Flow Cytometry*
Catalog # AATB22599
Supplier:  AAT Bioquest
CAS Number:  
undefined
Live or Dead™ Fixable Dead Cell Staining Kit *Red Fluorescence Optimized for Flow Cytometry*
Catalog # AATB22599
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Description:
    Live or Dead™ Fixable dead cell staining kit *Red fluorescence optimized for flow cytometry*
  • Tests per kit:
    200 Tests

Specifications

About this item

Our Live or Dead™ Fixable Dead Cell Staining Kits are a set of tools for labeling cells for fluorescence microscopic investigations of cellular functions.

  • Contains all necessary components
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.