ContactUsLinkComponent
PhosphoWorks™ Colorimetric ATP Assay Kit
PhosphoWorks™ Colorimetric ATP Assay Kit
Catalog # AATB21617
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
PhosphoWorks™ Colorimetric ATP Assay Kit
Catalog # AATB21617
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    PhosphoWorks™ Colorimetric ATP assay kit
  • Tests per kit:
    100 Tests
Specifications
About this item
Adenosine triphosphate (ATP) plays a fundamental role in cellular energetics, metabolic regulation and cellular signaling.
  • Simple and robust assay
  • Sensitive ATP detection
  • Multiple applications
Caution: For research use only.