Order Entry
Belgium
ContactUsLinkComponent
Cell Meter™ No Wash and Probenecid-Free Endpoint Calcium Assay Kit *Optimized for microplate reader*
Cell Meter™ No Wash and Probenecid-Free Endpoint Calcium Assay Kit *Optimized for microplate reader*
Catalog # AATB36312
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ No Wash and Probenecid-Free Endpoint Calcium Assay Kit *Optimized for microplate reader*
Catalog # AATB36312
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ no wash and probenecid-free endpoint calcium assay kit *optimized for microplate reader*
  • Tests per kit:
    100 Tests

Specifications

About this item

Cell Meter™ No Wash and Probenecid-Free Endpoint Calcium Assay Kit enables homogeneous fluorescence-based assays for detecting intracellular calcium mobilization without the need to use kinetics reading mode.

  • Ready to use
  • Contains all necessary components
  • Robust and adaptable
  • Optimized for microplate readers but suitable for all major fluorescence platforms
  • Multiple research applications
Caution: For research use only.