Order Entry
Belgium
ContactUsLinkComponent
Cell Meter™ Cellular Senescence Activity Assay Kit *Red Fluorescence*
Cell Meter™ Cellular Senescence Activity Assay Kit *Red Fluorescence*
Catalog # AATB23007
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Cellular Senescence Activity Assay Kit *Red Fluorescence*
Catalog # AATB23007
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    100 Tests
  • Description:
    Cell Meter™ cellular senescence activity assay kit *red fluorescence*
  • Tests per kit:
    100 Tests

Specifications

About this item

Cellular Senescence is an irreversible growth arrest triggered in order to prevent growth in DNA damaged cells.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.