Order Entry
Belgium
ContactUsLinkComponent
Cell Meter™ Autophagy Fluorescence Imaging Kit
Cell Meter™ Autophagy Fluorescence Imaging Kit
Catalog # AATB23001
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Autophagy Fluorescence Imaging Kit
Catalog # AATB23001
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Description:
    Cell Meter™ autophagy fluorescence Imaging kit
  • Tests per kit:
    200 Tests

Specifications

About this item

Autophagy is one of the major pathways for degradation of intracellular macromolecules in animal cells.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.