ContactUsLinkComponent
Cell Meter™ Live Cell Caspase 9 Binding Assay Kit *Green Fluorescence*
Cell Meter™ Live Cell Caspase 9 Binding Assay Kit *Green Fluorescence*
Catalog # AATB20117
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cell Meter™ Live Cell Caspase 9 Binding Assay Kit *Green Fluorescence*
Catalog # AATB20117
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    25 Tests
  • Description:
    Cell Meter™ Live cell Caspase 9 binding assay kit *Green fluorescence*
  • Tests per kit:
    25 Tests
Specifications
About this item
Our Cell Meter™ live cell caspases activity assay kits are based on fluorescent FMK inhibitors of caspases.
  • Based on FMK inhibitor
  • Direct caspase detection
  • Suitable for all major fluorescence platforms
Caution: For research use only.