DiTO™-3 [equivalent to TOTO®-3] *5 mM DMSO Solution* (0.2 ml)
Catalog # AATB17576
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:0,2 mL
- Description:DiTO™-3
- Purity:95%
Specifications
About this item
DiTO™-3 is chemically equivalent to TOTO®-3 (TOTO® is the trademark of Invitrogen).
- Cell-Impermeable
- Useful For Multicolor Imaging
- Bright Fluorescence
5 mM DMSO Solution
Caution: For research use only.