0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
iFluor®
iFluor® 488, Alkyne fluorescent probe
iFluor® 488, Alkyne fluorescent probe
iFluor® 488, Alkyne fluorescent probe
Catalog # AATB999
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
iFluor® 488, Alkyne fluorescent probe
Catalog # AATB999
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
Environmentally Preferable:
Specifications
About this item
Product Details
:
ADR:
5,II
:
GHS07
Product Details