6-HEX phosphoramidite for oligonucleotide labeling
Supplier: AAT BIOQUEST
Certificates
About this item
Synonyms: 5'-Hexachlorofluorescein phosphoramidite
CAS Number:1360547-55-2
:
Molecular Weight:1050.61 g/mol
ADR:5 II
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together